Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Aminohexan-1-ol
6-Aminohexan-1-ol for protein labeling
6-Aminohexan-1-ol for protein labeling
6-Aminohexan-1-ol for protein labeling
AATB4002
:
AAT BIOQUEST
:
6-Aminohexan-1-ol for protein labeling
AATB4002
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
100 g
Seq:
3401
:
:
117.19 g/mol
:
5,II
:
GHS07
:
4048-33-3
:
C₆H₁₅NO
:
MFCD00008241