Order Entry
Export
ContactUsLinkComponent
3-Cyanoumbelliferone ≥95%
Irritant
3-Cyanoumbelliferone ≥95%
Catalog # AATB40
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
3-Cyanoumbelliferone ≥95%
Catalog # AATB40
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :187.15 g/mol
ADR:5,II
 :GHS07
CAS Number:19088-73-4
Formula:C₁₀H₅NO₃
MDL Number:MFCD00037480