0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Biotin acid
Biotin acid, ReadiView™ biodetection
Biotin acid, ReadiView™ biodetection
Irritant
Biotin acid, ReadiView™ biodetection
Catalog # AATB3050
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Biotin acid, ReadiView™ biodetection
Catalog # AATB3050
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details