Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biotin PEG3 amine
Biotin PEG3 amine biodetection
Biotin PEG3 amine biodetection
Irritant
Biotin PEG3 amine biodetection
Catalog # AATB3024
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Biotin PEG3 amine biodetection
Catalog # AATB3024
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
ADR:
5,II
:
560.63 g/mol
:
GHS07
CAS Number:
194920-43-9
Product Details