Order Entry
Export
Orders LinkContactUsLinkComponent
Biotin NTA biodetection

Biotin NTA biodetection

Plastic bottle, 5 mg
Catalog # AATB3008
Irritant
Supplier:   AAT Bioquest
Biotin NTA biodetection
Catalog # AATB3008
Supplier:  AAT Bioquest
CAS Number:  176046-06-3
Biotin NTA biodetection
Biotin NTA biodetection

Plastic bottle, 5 mg

CAS Number:  176046-06-3
Price
142,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Biotin NTA (BNTA), a biotin-labeled nitrilotriacetic acid, is widely used to detect histidine-tagged proteins immobilized on nitrocellulose membranes with the detection limite of ~0.1 pmol.

  • Histidine tagging
Synonyms:  BNTA
  • CAS Number:176046-06-3
  • Molecular Weight:488.56 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg

Specifications