Order Entry
Export
ContactUsLinkComponent
 
 
Biotin-4-fluorescein ≥95%
  AATB3006
 :  
undefined
 
Biotin-4-fluorescein ≥95%
  AATB3006
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

 

 

 :
 :644.7 g/mol
 :5,II
 :GHS07
 :1032732-74-3
 :C₃₃H₃₂N₄O₈S