Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thioflavin T acid
Thioflavin T acid ≥90%
Thioflavin T acid ≥90%
Thioflavin T acid ≥90%
AATB23064
:
AAT Bioquest
:
Thioflavin T acid ≥90%
AATB23064
:
AAT Bioquest
:
Pk:
25 mg
Seq:
2315
:
:
496.55 g/mol