Order Entry
Export
ContactUsLinkComponent
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95%
Irritant
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95%
Catalog # AATB22221
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95%
Catalog # AATB22221
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :500.93 g/mol
ADR:5,II
 :GHS07
CAS Number:115532-50-8
Formula:C₂₅H₂₅N₂O₃·ClO₄
MDL Number:MFCD00274460