Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tide Quencher™ 3 amine
Tide Quencher™ 3 amine, Tide Quencher™ fluorescent probe
Tide Quencher™ 3 amine, Tide Quencher™ fluorescent probe
Tide Quencher™ 3 amine, Tide Quencher™ fluorescent probe
AATB2222
:
AAT Bioquest
:
Tide Quencher™ 3 amine, Tide Quencher™ fluorescent probe
AATB2222
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
609.62 g/mol
:
5,II
:
GHS07