Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
JC-1 iodide salt
JC-1 iodide salt ≥95%
JC-1 iodide salt ≥95%
:
AAT Bioquest
JC-1 iodide salt ≥95%
:
AAT Bioquest
:
47729-63-5
:
3520-43-2
:
C₂₅H₂₇Cl₄IN₄
:
MFCD00188475
:
652.23 g/mol
:
5,II
:
GHS07