Order Entry
Export
ContactUsLinkComponent
DiOC₅(3) iodide ≥95%
DiOC₅(3) iodide ≥95%
Catalog # AATB22045
Supplier:  AAT Bioquest
CAS Number:  
undefined
DiOC₅(3) iodide ≥95%
Catalog # AATB22045
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :544.47 g/mol
CAS Number:53213-81-3
Formula:C₂₇H₃₃IN₂O₂