Order Entry
Export
ContactUsLinkComponent
 
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥95%
  AATB22038
 :  
undefined
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥95%
  AATB22038
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :460.32 g/mol
 :905-96-4
 :C₂₁H₂₁IN₂O₂
 :MFCD00011952