Order Entry
Export
ContactUsLinkComponent
 
Irritant
Cal-520N™ potassium salt
Catalog # AATB21147
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Cal-520N™ potassium salt
Catalog # AATB21147
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :1004.17 g/mol
ADR:5,II
 :GHS07