Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluo-8L™ sodium salt
Fluo-8L™ sodium salt
Fluo-8L™ sodium salt
Irritant
Fluo-8L™ sodium salt
Supplier:
AAT BIOQUEST
Certificates
Expand image
Irritant
Fluo-8L™ sodium salt
Supplier:
AAT BIOQUEST
About this item
:
ADR:
5,II
:
828.54 g/mol
:
GHS07