Order Entry
Export
ContactUsLinkComponent
 

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, EGTA tetrasodium
 :
 :468.28 g/mol
ADR:5,II
 :GHS07
CAS Number:13368-13-3
Formula:C₁₄H₂₀N₂Na₄O₁₀
MDL Number:MFCD00274420