Order Entry
Export
ContactUsLinkComponent
Tide Quencher™ 5WS alkyne, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 5WS alkyne, Tide Quencher™ fluorescent probe
Catalog # AATB2083
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Tide Quencher™ 5WS alkyne, Tide Quencher™ fluorescent probe
Catalog # AATB2083
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :927.06 g/mol
ADR:5,II
 :GHS07