
Trehalose hexaacetate cell permeable, CytoWatch™
Catalog # AATB20040
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:594.52 g/mol
ADR:5,II
:GHS07
CAS Number:67398-72-5