Order Entry
Export
ContactUsLinkComponent
 
Cy5.5NS succinimidyl ester ≥90%
Catalog # AATB199
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cy5.5NS succinimidyl ester ≥90%
Catalog # AATB199
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :730.33 g/mol