Order Entry
Export
ContactUsLinkComponent
XFD488 C5 Maleimide fluorescent probe
XFD488 C5 Maleimide fluorescent probe
Catalog # AATB1878
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
XFD488 C5 Maleimide fluorescent probe
Catalog # AATB1878
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :720.66 g/mol
ADR:5,II
 :GHS07