Order Entry
Export
ContactUsLinkComponent
TWO-PRO™ 1 5 mM in DMSO ≥95%
Irritant
TWO-PRO™ 1 5 mM in DMSO ≥95%
Catalog # AATB17571
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
TWO-PRO™ 1 5 mM in DMSO ≥95%
Catalog # AATB17571
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    0,2 mL
  • Seq:
    2020

Specifications

About this item

 :
 :645.38 g/mol
ADR:5,II
 :GHS07
CAS Number:157199-59-2
Formula:C₂₄H₂₉I₂N₃S