
Hydroxystilbamidine bis(methanesulphonate) ≥95%
Catalog # AATB17514
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:472.54 g/mol
ADR:5,II
:GHS07
CAS Number:223769-64-0
Formula:C₁₈H₂₄N₄O₇S₂
MDL Number:MFCD01863083