
7-Aminoactinomycin D (7-AAD) ≥95%
Catalog # AATB17501
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:1270.43 g/mol
ADR:5,II
:GHS07
CAS Number:7240-37-1
Formula:C₆₂H₈₇N₁₃O₁₆
MDL Number:MFCD00077295