Order Entry
Export
ContactUsLinkComponent
Cyanine 5.5 bisacid ≥95% [equivalent to Cy5.5® bisacid]
Cyanine 5.5 bisacid ≥95% [equivalent to Cy5.5® bisacid]
Catalog # AATB172
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cyanine 5.5 bisacid ≥95% [equivalent to Cy5.5® bisacid]
Catalog # AATB172
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :1117.17 g/mol
 :146368-10-7