7-Deaza-7-Propargylamino-3'-azidomethyl-dATP
Catalog # AATB17090
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 Nmo
- Seq:9030
Specifications
About this item
:
:598.3 g/mol
ADR:5,II
:GHS07
CAS Number: 666847-93-4
Formula:C15H21N8O12P3