0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Aminopropargyl dCTP
Aminopropargyl dCTP for PCR
Aminopropargyl dCTP for PCR
AATB17050
:
AAT Bioquest
:
Aminopropargyl dCTP for PCR
AATB17050
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
10 µmol
Seq:
9105
:
:
543.95 g/mol
:
5,II
:
GHS07
:
2088422-92-6