Order Entry
Export
ContactUsLinkComponent
 
Z-VAD(OMe)-FMK
  AATB13300
 :  
undefined
Z-VAD(OMe)-FMK
  AATB13300
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • CAS No.:
    187389-52-2
  • MDL No.:
    MFCD02684037
  • Protein synonyms:
    Carbobenzoxy-valyl-alanyl-aspartyl-[O-methyl]- fluoromethylketone|N-Benzyloxycarbonyl-Val-Ala-Asp(O-Me) fluoromethyl ketone
  • Protein/peptide name:
    Z-VAD(OMe)-FMK
  • Molecular weight:
    467.48 g/mol
  • Molecular formula:
    C₂₂H₃₀FN₃O₇

 

 

Z-VAD-FMK (carbobenzoxy-valyl-alanyl-aspartyl-[O-methyl]- fluoromethylketone) is a cell-permeable, irreversible pan caspase inhibitor that binds to the catalytic site of caspase proteases and can inhibit induction of apoptosis.

  • Caspase and apoptosis inhibitor
  • Enhanced stability
  • Increased cell permeability

For: Molecular biology, Labeling, Proteomics

GHS Symbol: GHS07
Hazard Class 5
Packing Group II
Signalword: Warning