Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-Luciferin-6-O-β-D-galactopyranoside
D-Luciferin-6-O-β-D-galactopyranoside ≥95%
D-Luciferin-6-O-β-D-galactopyranoside ≥95%
AATB12513
:
AAT BIOQUEST
:
D-Luciferin-6-O-β-D-galactopyranoside ≥95%
AATB12513
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
400.427 g/mol
:
5,II
:
GHS07
:
131474-38-9
:
C₂₂H₂₄O₇
:
MFCD09038345