iFluor® 555, Hydrazide
Plastic bottle, 1 mg
Catalog # AATB1083
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright and photostable
- Minimal quenching
- Stable conjugation
- pH independent fluorescence between 3-11
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1 mg



