iFluor® 488 fluorescent probe
Plastic bottle, 1 mg
Catalog # AATB1014
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.
- pH-insensitive
- Azide reactive
- High photostability
- Brighter signal
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1 mg



