Order Entry
Export
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 488)
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 488)
Supplier:  AAT Bioquest
Certificates
undefined
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 488)
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • AATB16828
  • AATB16678
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • iFluor® 488
  • iFluor® 488
  • Clone:
  • IgG
  • IgG
  • Reactivity:
  • Rabbit
  • Rabbit
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)
  • Cross adsorption:
  • Min×Human
  • Min×Human

Specifications