Order Entry
Export
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Agarose)
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Agarose)
Catalog # AATB55010
Supplier:  AAT BIOQUEST
undefined
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Agarose)
Catalog # AATB55010
Supplier:  AAT BIOQUEST

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Clone:
    IgG
  • Conjugation:
    Agarose
  • Host:
    Goat
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Mouse
  • Pack type:
    Vial
  • Pk:
    10 mg

Specifications

About this item

Agarose conjugated antibodies are designed for the rapid and efficient collection of antibodies and proteins, including recombinant fusion proteins, from a complex protein mixture (see protocol).

  • Excellent quality
  • Affinity gel building block
  • Multiple applications
Caution: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse