Streptavidin
Supplier: AAT BIOQUEST
Certificates
About this item
Streptavidin is a biotin-binding protein found in the culture broth of the bacterium Streptomyces avidinii.
- Extremely high biotin-binding affinity
- High quality
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin, Streptomyces avidinii
- Protein/peptide name:Streptavidin