Order Entry
Export
ContactUsLinkComponent
Native Streptavidin (from Streptomyces avidinii), HRP (Horseradish Peroxidase)
Native Streptavidin (from Streptomyces avidinii), HRP (Horseradish Peroxidase)
Catalog # AATB16920
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Native Streptavidin (from Streptomyces avidinii), HRP (Horseradish Peroxidase)
Catalog # AATB16920
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin| Streptomyces avidinii

Specifications

About this item

Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.

  • Extremely useful conjugate
  • Multiple applications
  • In-house quality control
Caution: For research use only.