Order Entry
Export
ContactUsLinkComponent
Streptavidin, iFluor® 750
Streptavidin, iFluor® 750
Supplier:  AAT BIOQUEST
Certificates
undefined
Streptavidin, iFluor® 750
Supplier:  AAT BIOQUEST

Specifications

  • Conjugation:
    iFluor® 750
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii

Specifications