Order Entry
Export
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Violet 450
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Violet 450
Catalog # AATB16930
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Violet 450
Catalog # AATB16930
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 µG
  • Conjugation:
    mFluor™ Violet 450
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin
  • Protein/peptide name:
    Streptavidin

Specifications

About this item

Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.

  • Optimized for flow cytometry
  • Useful for assay development
  • High affinity
  • Multiple applications
Caution: For research use only.