Native Bovine BSA (from Serum), Estradiol
AATB50552
:
- Pack type:Vial
- Pk:5 mg
- Conjugation:Estradiol
- Protein/peptide type:Native
- Source:Serum
- Species:Bovine
- Protein synonyms:Bovine Serum Albumin
- Protein/peptide name:BSA
Estradiol is an estrogen steroid hormone and the major female sex hormone.
- Assay development
- High sensitivity
: For research use only.