Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 594
Catalog # AATB23122
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:300 Tests
- Conjugation:Alexa Fluor® 594
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Species:Amanita phalloides
- Protein synonyms:Amanita phalloides
- Protein/peptide name:Phalloidin
Specifications
About this item
This red fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 594-labeled phalloidin) selectively binds to F-actins.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Caution: For research use only.