Order Entry
Export
ContactUsLinkComponent
Nuclear Violet™ LCS1
Nuclear Violet™ LCS1
Catalog # AATB17543
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Nuclear Violet™ LCS1
Catalog # AATB17543
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    0,5 mL
  • Description:
    Nuclear Blue™ LCS1
  • Purity:
    95%

Specifications

About this item

Our Nuclear Violet™ LCS1 is a fluorogenic, DNA-selective and cell-permeant dye for analysing DNA content in living cells.

  • Cell-impermeable
  • Useful for multicolour imaging
  • Bright fluorescence

5 mM DMSO solution.

Caution: For research use only.