LysoBrite™ Green
Catalog # AATB22643
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:500 Tests
- Description:LysoBrite™ green
- Purity:90%
Specifications
About this item
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Lysosome-specific dye
- Long-term use
- Low cytotoxicity
Caution: For research use only.