ContactUsLinkComponent
CytoFix™ Red Lysosomal Stain
  AATB23210
 :  
undefined
CytoFix™ Red Lysosomal Stain
  AATB23210
 :  
  • Pk:
    500 Tests
  • Description:
    CytoFix™ red lysosomal stain
  • Purity:
    95%
  • Tests per kit:
    500 Tests
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste and cellular debris through a process known as autophagy.
  • High Staining Efficiency
  • Lysosome-Specific Dye
  • Long-Term Cell Tracking
  • High Efficiency
 : For research use only.