CellPaint™ TSP membrane stain
Catalog # AATB22700
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:500 Tests
- Description:TSP
- Purity:90%
- Tests per kit:500 Tests
Specifications
About this item
TSP is a styrylpyridine-based fluorescent membrane probe suitable for imaging plasma membranes in living cells and tissues.
- Low Cytotoxicity
- High Target Affinity
- Bright Fluorescence
Caution: For research use only.