Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulfide Green™ AM
Sulfide Green™ AM
Sulfide Green™ AM
Sulfide Green™ AM
Catalog # AATB21301
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Sulfide Green™ AM
Catalog # AATB21301
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
500 µG
Seq:
1418
Specifications
About this item
Product Details
:
Product Details