Ac-IETD-AFC
Catalog # AATB13410
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- CAS No.:211990-57-7
- Protein/peptide name:Ac-IETD-AFC
- Purity:95%
Specifications
About this item
Ac-IETD-AFC is a fluorogenic caspase-8/granzyme B substrate containing the acetyl (Ac) moiety.
- Hazard Class/ Packing Group: 5, II
- GHS symbol: GHS07
- Signal Word: Warning