Order Entry
Export
ContactUsLinkComponent
Ac-DEVD-CHO
Ac-DEVD-CHO
Catalog # AATB13403
Supplier:  AAT Bioquest
CAS Number:  
undefined
Ac-DEVD-CHO
Catalog # AATB13403
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • CAS No.:
    169332-60-9
  • Protein/peptide name:
    Ac-DEVD-CHO
  • Purity:
    90%

Specifications

About this item

Ac-DEVD-CHO is a reversible inhibitor of IL-1ß converting enzyme (ICE); inhibits poly(ADP-ribose) polymerase cleavage by apopain (caspase 3).

  • Hazard Class/ Packing Group: 5, II
  • GHS symbol: GHS07
  • Signal Word: Warning