Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
JC-10
JC-10
JC-10
JC-10
Catalog # AATB22204
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
JC-10
Catalog # AATB22204
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic vial
Pk:
500 µl
Seq:
1418
Pk Info:
500 tests (5×100 µl)
Specifications
About this item
Product Details
:
:
583.34 g/mol
Product Details