CytoTrace™ Green CMFDA
Catalog # AATB22017
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:1 mg
- Description:CytoTrace™ green CMFDA
- Purity:95%
Specifications
About this item
CytoTrace™ Green CMFDA is an excellent replacement to CellTracker™ Green CMFDA (Invitrogen C2925, CellTracker™ is the trademark of Invitrogen).
- Robust
- High Multigenerational Cell Retention
- Bright Signal
Caution: For research use only.