Order Entry
Export
ContactUsLinkComponent
CytoTrace™ Red CMTPX
CytoTrace™ Red CMTPX
Catalog # AATB22015
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
CytoTrace™ Red CMTPX
Catalog # AATB22015
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Description:
    CytoTrace™ red CMTPX
  • Purity:
    85%

Specifications

About this item

CytoTrace™ Red CMTPX is chemically same to CellTracker™CMTPX.

  • High Multigenerational Cell Retention
  • Bright Signal
  • Identical To CellTracker™CMTPX
Caution: For research use only.