Orders LinkContactUsLinkComponent
RatioWorks™ PH165
500 µG
  AATB21213
RatioWorks™ PH165
  AATB21213
 :  
RatioWorks™ PH165
RatioWorks™ PH165
500 µG
 :  
224,00 €
1 * 500 µG
RatioWorks™ PH165 is a novel fluorescent probe that can be used to measure intracellular pH in live cells.
  • pH-dependent fluorescence
  • Ratiometric detection
  • Long-wavelength emission
  • Cell impermeable
 : For research use only.
  • Pk:
    500 µG