Order Entry
Export
ContactUsLinkComponent
 
Cell Meter™ Autophagy Assay Kit *Green Fluorescence*
Catalog # AATB23002
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Autophagy Assay Kit *Green Fluorescence*
Catalog # AATB23002
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ autophagy assay kit *green fluorescence*

Specifications

About this item

This Cell Meter™ autophagy kit employs autophagy Green™ as a specific autophagosome marker to analyse the activity of autophagy.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.