Total Residual Chlorine Standard Certified Reference Material
Catalog # ACCUWC-TRC-10X-10M
Supplier: AccuStandard
CAS Number:
Specifications
- Pk:100 mL
- Description:Total residual chlorine standard certified reference material
- Environmentally Preferable:
- Storage temperature:0...5 °C